You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
New Products
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Home
▸
Shop All Products
▸
Protein Analysis Reagents
▸
Amino Acid Analysis Reagents and Accessories
▸
HOBt/DMF (0.5 M 1-Hydroxybenzotriazole/N, N-Dimethylformamide), 200 mL
Applied Biosystems™
HOBt/DMF (0.5 M 1-Hydroxybenzotriazole/N, N-Dimethylformamide), 200 mL
This is a reagent for use in peptide synthesis.For Research Use Only. Not for use in diagnostics
procedures.
Read more
Have Questions?
Catalog Number
Quantity
400934
200 mL
Catalog number
400934
Price (MXN)
-
Quantity:
200 mL
This is a reagent for use in peptide synthesis.
For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)
43X Peptide Synthesizer
Quantity
200 mL
Reagent Type
Peptide Synthesis Coupling Reagent
Shipping Condition
Room Temperature
Form
Liquid
Product Type
Peptide Synthesis Reagent
Unit Size
Each
Contents & Storage
Store at 4°C.