β-Lactoglobulin - FAQs
β-Lactoglobulin - FAQs
View additional product information for β-Lactoglobulin - FAQs (400979)
1 product FAQs found
What is the extended sequence of Beta-lactoglobulin?
For the first 40 residues, the sequence is as shown below:
LIVTQ - TMKGL - DIQKV - AGTWY - SLAMA - ASDIS - LLDAQ-SAPLR
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.