Applied Biosystems™

GeneScan™ 2500 ROX™ dye Size Standard

Catalog number:  401100

 Related applications:

Sanger Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The GeneScan™ 2500 ROX™ Size Standard is a ROX™ dye-labeled size standard that is designed for reproducible sizing of fragment analysis data from non-denaturing assays. Use this standard for fragments between 55 and 2500 bp. The standard contains 27 ROX™ dye-labeled DNA fragments.

For Research Use Only. Not for use in diagnostics procedures.

Specifications

For Use With (Equipment): 310 Genetic Analyzer, 3130xl Genetic Analyzer, 3500 Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer, 3730xl DNA Analyzer
Label or Dye: ROX
Product Line: GeneScan™, ROX™
Product Size: 400 reactions
Research Category: Signal Transduction
Shipping Condition: Wet Ice

Contents & storage

The kit contains four tubes of standard (50 µL in each tube), sufficient for a total of 400 analyses. The kit may be stored up to one year at 2°C to
4°C. (Do not freeze)

Documents

Manuals & protocols