Dye Primer/Matrix Standards [5FAM™, JOE™, TAMRA™, and ROX™ dyes]

Catalog number: 401114

Applied Biosystems™  Related applications: Genotyping & Genomic Profiling | Sanger Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This Dye Primer Matrix Standards are used to generate the multicomponent matrix required when analyzing 5-FAM™, JOE™, TAMRA™ and ROX™-labeled DNA fragments. The standard can be used for Dye Primer cycle Sequencing and Dye Primer Sequenase Sequencing and on the Applied Biosystems® 310, 3100⁄3100-Avant™ and 3130⁄XL Genetic Analyzers. The Data Collection Software for these instruments uses the multicomponent matrix to automatically analyze the four different colored fluorescent dye-labeled samples in a single capillary.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): 310 Genetic Analyzer
Label or Dye: FAM™, JOE™, ROX™, TAMRA™
Product Line: Applied Biosystems™
Product Size: 1 each
Shipping Condition: Wet Ice

Contents & storage

Contains dye labeled standards used to generate multicomponent matrix. 1 tube of each dye. Store the kit at 2 - 8°C. Do not freeze.

Documents

Manuals & protocols