Ammonium persulfate, 98+%, ACS reagent
Ammonium persulfate, 98+%, ACS reagent
Ammonium persulfate, 98+%, ACS reagent
Ammonium persulfate, 98+%, ACS reagent
Thermo Scientific Chemicals

Ammonium persulfate, 98+%, ACS reagent

Oxidation reagent | CAS: 7727-54-0 | H8N2O8S2 | 228.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
500 g
2 kg
5 kg
25 kg
Packaging:
View all
Fibre drum
Catalog number 401160250
Price (EUR)
878,00
Each
Quantity:
25 kg
Packaging:
Fibre drum
Request bulk or custom format
Price (EUR)
878,00
Each
Chemical Identifiers
CAS7727-54-0
IUPAC Namediammonium [(sulfonatoperoxy)sulfonyl]oxidanide
Molecular FormulaH8N2O8S2
InChI KeyROOXNKNUYICQNP-UHFFFAOYSA-N
SMILES[NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals
Titration with KMnO4>=98.0 %
Heavy metals (as Pb)=<0.005 %
Residue after ignition=<0.05 %
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Oxidation reagent
RUO – Research Use Only