8-Anilino-1-naphthalenesulfonic acid, 98%
8-Anilino-1-naphthalenesulfonic acid, 98%
8-Anilino-1-naphthalenesulfonic acid, 98%
Thermo Scientific Chemicals

8-Anilino-1-naphthalenesulfonic acid, 98%

CAS: 82-76-8 | C16H13NO3S | 299.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 401211000
Price (TWD)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS82-76-8
IUPAC Name8-(phenylamino)naphthalene-1-sulfonic acid
Molecular FormulaC16H13NO3S
InChI KeyFWEOQOXTVHGIFQ-UHFFFAOYSA-N
SMILESOS(=O)(=O)C1=C2C(NC3=CC=CC=C3)=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=96.0 %
Appearance (Color)Dark green to brown to black
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only