The 1.0 μmol dG(dmf) Controlled Pore Glass (CPG) columns are suitable for the synthesis of oligonucleotides<50 bases in length. These columns deliver high-quality oligonucleotides.
• Proprietary design and manufacturing methods ensure consistency and high quality • Each column is tested to verify accuracy and precision of fill, then pressure-tested to verify proper assembly and to guarantee each column is leak-free • The 500 angstrom pore size allows synthesis of oligonucleotides of up to 50 bases.
These columns are fully compatible with the Applied Biosystems 3400 instruments, ABI 391, ABI 392, ABI 394, and earlier instruments.
For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Column Type
Oligonucleotide Synthesis Column
Format
Column
Sample Type
Oligos
For Use With (Equipment)
3900 DNA Synthesizer
For Use With (Application)
Oligo Synthesis
Matrix
Controlled Pore Glass
Pore Size
500 Å
Prep Scale
1 μmol
Product Type
Oligonucleotide Synthesis Columns
Shipping Condition
Room Temperature
Target
Nucleic Acids
Contents & Storage
Columns may be stored in a dry environment at room temperature.