L(+)-Asparagine monohydrate, specified according the requirements of Ph.Eur.
L(+)-Asparagine monohydrate, specified according the requirements of Ph.Eur.
L(+)-Asparagine monohydrate, specified according the requirements of Ph.Eur.
Thermo Scientific Chemicals

L(+)-Asparagine monohydrate, specified according the requirements of Ph.Eur.

CAS: 5794-13-8 | C4H10N2O4 | 150.134 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
1 kg
Catalog number 401480010
Price (USD)
422.65
Online exclusive
469.00
Save 46.35
Each
Quantity:
1 kg
Request bulk or custom format
Price (USD)
422.65
Online exclusive
469.00
Save 46.35
Each
Chemical Identifiers
CAS5794-13-8
IUPAC Name(2S)-2-amino-3-carbamoylpropanoic acid hydrate
Molecular FormulaC4H10N2O4
InChI KeyRBMGJIZCEWRQES-DKWTVANSSA-N
SMILESO.N[C@@H](CC(N)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Chlorides=<200 ppm
Titration with HClO499.0 to 101.0 % (on dried substance)
Loss on drying10.5 % to 12.5 % (1 g, 130°C, 3 h)
Sulfated ash=<0.1 % (1 g)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only