Azelaic acid, 98%
Azelaic acid, 98%
Azelaic acid, 98%
Thermo Scientific Chemicals

Azelaic acid, 98%

CAS: 123-99-9 | C9H16O4 | 188.223 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
View all
Glass bottle
Catalog number 401520250
Price (USD)
28.70
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
28.70
Each
Chemical Identifiers
CAS123-99-9
IUPAC Namenonanedioic acid
Molecular FormulaC9H16O4
InChI KeyBDJRBEYXGGNYIS-UHFFFAOYSA-N
SMILESOC(=O)CCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder
GC>=97.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only