Applied Biosystems™

Reaction Vessel, 40 mL, RTF version

Catalog number:  401573

 Related applications: Protein Sequencing & Peptide Synthesis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

A new style reaction vessel is now available for both the Models 430A and 431A Peptide Synthesizers. This new vessel uses tabbed filters which are placed on the recessed edge inside the vessel. The caps of the new vessel include an internal O-ring to back up the primary seal formed between the vessel, filter, and the cap. These vessels require changing the side of the vortexer bracket to accommodate the vessel and include a shielded vortexer bearing. These design changes address the known difficulties associated with the assembly of the reaction vessels.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): 433A Peptide Synthesizer
Product Size: 40 mL
Shipping Condition: Room Temperature
Type: Reaction Vessel

Contents & storage

Store at room temperature.

Documents

Manuals & protocols