Procise™ Reagent Kit B, for 500 Cycles

Catalog number: 401712

Applied Biosystems™  Related applications: Protein Sequencing & Peptide Synthesis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This kit contains reagents for Procise® N-terminal protein sequencing.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): Procise Protein Sequencer
Form: Liquid
Product Line: Procise®
Product Size: 1 kit
Reagent Type: N-terminal Sequencing Reagent

Contents & storage

This kit contains the following reagents: Trifluoroacetic Acid, N-Butyl Chloride, 25% Trifluoroacetic Acid in Water, 5% PITC in N-Heptane, 20% Acetonitrile in Water, Ethyl Acetate, Premix Buffer Concentrate, 3.5% THF⁄Water, Isopropanol and Acetonitrile, N-Methylpiperidine in Isopropanol, Butanol and Water, ACN⁄AcetylCys, and PTH-AA Standard Solution. Each may be sold separately. Store individual components according to label.

Documents

Manuals & protocols