GeneScan™ 350 TAMRA™ dye Size Standard

Catalog number: 401736

Applied Biosystems™  Related applications: Genotyping & Genomic Profiling | Sanger Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The GeneScan™ 350 TAMRA™ Size Standard is a TAMRA™ dye-labeled size standard for the reproducible sizing of fragment analysis data.

GeneScan 350 TAMRA™ Size Standard is designed for sizing DNA fragments in the 35-350 nucleotides range and provides 12 single-stranded labeled fragment of: 35, 50, 75, 100, 139, 150, 160, 200, 250, 300, 340 and 350 nucleotides. Each of the DNA fragments is labeled with the TAMRA™ fluorophore which results in a single peak when run under denaturing conditions.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): 310 Genetic Analyzer, 3130xl Genetic Analyzer, 3500 Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer, 3730xl DNA Analyzer
Label or Dye: TAMRA™
Product Line: GeneScan™, TAMRA™
Product Size: 800 reactions
Shipping Condition: Wet Ice

Contents & storage

Contains12 TAMRA labeled single stranded DNA fragments for use as a standard. Store the kit at 2-8°C. (Do not freeze).

Documents

Manuals & protocols