TFA Treated Cartridge Filters (6mm) for Procise™ cLC System - FAQs
TFA Treated Cartridge Filters (6mm) for Procise™ cLC System - FAQs
View additional product information for TFA Treated Cartridge Filters (6mm) for Procise™ cLC System - FAQs (401870)
1 product FAQs found
Can I sequence an FMOC-peptide that is still resin-bound?
Yes. Apply a few beads to the Biobrene-treated glass fiber filter and sequence as you normally would. Be sure the peptide is deprotected, otherwise the Fmoc group serves to N-terminally block it. The TFA in the Edman chemistry should deprotect most (though perhaps not all) side-chain groups, and will cleave the peptide from the resin (though the latter is not necessary).