310 Genetic Analyzer Buffer Vials, 4 mL

Catalog number: 401955

Applied Biosystems™  Related applications: Sanger Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

These vials for the 310 Genetic Analyzer autosampler hold buffer and water. Cap adapters are included.

Use with the 310 Genetic Analyzer with Windows® OS, and septa for 0.5 ml sample tubes.

For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): 310 Genetic Analyzer
Product Line: Applied Biosystems™
Product Size: 50 vials
Shipping Condition: Room Temperature
Type: Containers & Vessels

Contents & storage

Each package contains 50 vials and cap adapters. May be stored at ambient temperature.

Documents

Manuals & protocols