Applied Biosystems™

Septa for 0.5 mL sample tubes (for the 310 Genetic Analyzer)

Catalog number:  401956

Related applications:

Sequencing

Back to top

Description

These are septa covers for 0.5 ml sample tubes, for use with the 310 Genetic Analyzer.

Use with: 1.0 mL Glass Syringe, 310 Genetic Analyzer Capillary (47 cm, 36 cm wtr), 310 Running Buffer (10X), BigDye® Terminator v1.1 Cycle Sequencing Kit, GeneScan™ 500 ROX™ Size Standard, Genetic Analyzer Sample Tubes, 0.5 ml, Hi-Di™ Formamide, MicroAmp® Reaction Tube with Cap (0.2 ml), POP-4™ Polymer for the 310 Genetic Analyzer, POP-6™ Polymer for the 310 Genetic Analyzer, Quantifiler® Human DNA Quantification Kit.

For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): 310 Genetic Analyzer
Shipping Condition: Room Temperature
Tube Type: Disposable Tube, Conical Tube, Assay Tube

Contents & storage

Each package contains 500 septa.