Thermo Scientific Chemicals

Benzoyl fluoride, 97%, Thermo Scientific Chemicals

Catalog number: 402040250
25 g, Each
Thermo Scientific Chemicals

Benzoyl fluoride, 97%, Thermo Scientific Chemicals

Catalog number: 402040250
25 g, Each
Quantity

Chemical Identifiers

CAS
455-32-3
Appearance (Color)
Clear colorless to yellow to brown
Infrared spectrum
Conforms
GC
>=96.0 %
Appearance (Form)
Liquid
Refractive index
1.4960 to 1.4990 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text