Bis(2-hydroxyethyl)dimethylammonium chloride, 99%
Bis(2-hydroxyethyl)dimethylammonium chloride, 99%
Bis(2-hydroxyethyl)dimethylammonium chloride, 99%
Thermo Scientific Chemicals

Bis(2-hydroxyethyl)dimethylammonium chloride, 99%

CAS: 38402-02-7 | C6H16ClNO2 | 169.65 g/mol
Have Questions?
Catalog NumberQuantityPackaging
40253025025 gGlass bottle
Catalog number 402530250
Price (EUR)
90,65
Online exclusive
101,00
Save 10,35
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
90,65
Online exclusive
101,00
Save 10,35
Each
Chemical Identifiers
CAS38402-02-7
IUPAC Namebis(2-hydroxyethyl)dimethylazanium chloride
Molecular FormulaC6H16ClNO2
InChI KeyOPVXPOIGBSHXNM-UHFFFAOYSA-M
SMILES[Cl-].C[N+](C)(CCO)CCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white to off-white
Titration Argentometric>=98.5 %
Appearance (Form)Adhering crystalline powder or crystals
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only