Borane-trimethylamine complex, 97%
Borane-trimethylamine complex, 97%
Borane-trimethylamine complex, 97%
Thermo Scientific Chemicals

Borane-trimethylamine complex, 97%

CAS: 75-22-9 | C3H12BN | 72.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 402740250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS75-22-9
IUPAC Nametrimethylamine borane
Molecular FormulaC3H12BN
InChI KeyWVMHLYQJPRXKLC-UHFFFAOYSA-N
SMILESB.CN(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder or crystals and/or chunks
Infrared spectrumConforms
Titration with Na2S2O3>=96.0 %
Melting point92°C to 96°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only