Bromelain, sourced from stem, >=1200 GDU/g (>=2.4 FIP U/mg)
Thermo Scientific Chemicals

Bromelain, sourced from stem, >=1200 GDU/g (>=2.4 FIP U/mg)

CAS: 37189-34-7
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 402830250
Price (USD)
61.65
Special offer
Online exclusive
Ends: 28-Dec-2026
72.00
Save 10.35
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
61.65
Special offer
Online exclusive
Ends: 28-Dec-2026
72.00
Save 10.35
Each
Chemical Identifiers
CAS37189-34-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Specific activity>=1200 GDU/g
Residue after ignition=<5 %
Appearance (Color)Cream to light brown
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only