Bromine, 99+%, extra pure
Bromine, 99+%, extra pure
Bromine, 99+%, extra pure
Thermo Scientific Chemicals

Bromine, 99+%, extra pure

CAS: 7726-95-6 | Br | 79.91 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
100 mL
500 mL
1 L
Packaging:
Plastic Coated Glass Bottle
產品號碼 402841000
價格 (TWD)
-
Quantity:
100 mL
Packaging:
Plastic Coated Glass Bottle
Request bulk or custom format
化學識別
CAS7726-95-6
IUPAC Namebromide
Molecular FormulaBr
InChI KeyCPELXLSAUQHCOX-UHFFFAOYSA-M
SMILES[Br-]
檢視更多
規格Specification Sheet規格表
Titration Iodometric>=99.0 %
Iodine (I)=<10 ppm
Appearance (Color)Red-brown
Heavy metals (as Pb)=<2 ppm
Chlorine (Cl)=<500 ppm
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only