Applied Biosystems™

310 Genetic Analyzer Septa Retainer Clips for 96-Tube Sample Tray

Catalog number:  402866

 Related applications: Sanger Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

These are septa retainer clips for the 96-well tray assembly of the 310 Genetic Analyzer.

For use with: 96-Well Tray Adapter, MicroAmp® 8-Tube Strip (0.2 ml), MicroAmp® 96-Well Tray⁄Retainer Set, MicroAmp® Splash Free 96-Well Base, and Septa for 0.2 ml MicroAmp™ Tubes

For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): 310 Genetic Analyzer
Plate Compatibility: 96-Well Plates
Product Size: 1 package
Shipping Condition: Room Temperature
Type: Retainer Clip

Contents & storage

Each package contains 4 retainer clips.

Documents

Manuals & protocols