3-Bromo-2-butanone, 95%, stabilized
3-Bromo-2-butanone, 95%, stabilized
3-Bromo-2-butanone, 95%, stabilized
Thermo Scientific Chemicals

3-Bromo-2-butanone, 95%, stabilized

CAS: 814-75-5 | C4H7BrO | 151 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 402910050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS814-75-5
IUPAC Name3-bromobutan-2-one
Molecular FormulaC4H7BrO
InChI KeyBNBOUFHCTIFWHN-UHFFFAOYNA-N
SMILESCC(Br)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=94 %
Appearance (Color)Clear light yellow to brown
Infrared spectrumConforms
Stabilizer=<1 % magnesium oxide
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only