Bromophenol Blue, pure, 0.1% in water
Bromophenol Blue, pure, 0.1% in water
Bromophenol Blue, pure, 0.1% in water
Thermo Scientific Chemicals

Bromophenol Blue, pure, 0.1% in water

CAS: 115-39-9 | C19H10Br4O5S | 669.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 403150250
Price (EUR)
61,65
Online Exclusive
68,10
Save 6,45 (9%)
Each
Quantity:
25 mL
Request bulk or custom format
Price (EUR)
61,65
Online Exclusive
68,10
Save 6,45 (9%)
Each
Chemical Identifiers
CAS115-39-9
IUPAC Name3,3-bis(3,5-dibromo-4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Molecular FormulaC19H10Br4O5S
InChI KeyUDSAIICHUKSCKT-UHFFFAOYSA-N
SMILESOC1=C(Br)C=C(C=C1Br)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Br)=C(O)C(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Concentration(by UV) About 0.1%
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only