Contains 100 mg TET™ Dye Phosphoramidite.TET™ Dye Phosphoramidite is used for the 5’ attachment of TET™ dye during oligonucleotide synthesis.TheRead more
Have Questions?
Change view
Catalog Number
Quantity
403171
50 mg
401533
100 mg
2 Options
Catalog number 403171
Price (USD)
494.00
Each
Quantity:
50 mg
Price (USD)
494.00
Each
Contains 100 mg TET™ Dye Phosphoramidite.
TET™ Dye Phosphoramidite is used for the 5’ attachment of TET™ dye during oligonucleotide synthesis.
The label is treated as if it were simply another base: It is inserted at the appropriate place in the sequence and incorporated automatically.
• Specialty phosphoramidites allow online addition of the label during synthesis, saving time in production of labeled probes and primers. • Designed and optimized for use with ABI synthesizers, guaranteeing reliable nucleic acid synthesis. • We offer a range of specialty phosphoramites and linkers to give you the flexibility demanded in oligonucleotide synthesis.
On-instrument labeling typically saves hours of post-synthesis manual processing Fluorescent label TET™ is a single-isomer fluorescein dye designed for detection and analysis of labeled PCR products on ABI PRISM™ genetic analysis systems. TET™ is equally useful in applications historically employing isotopic labels. As such, it eliminates the hazard associated with handling, storage, and disposal of radioactive materials.
Note: See user's manual or package insert for limited label license, and trademark information. For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)ABI Synthesizers, Guaranteeing Reliable Nucleic Acid Synthesis
For Use With (Equipment)3900 DNA Synthesizer
FormatPowder
Nucleic Acid TypeNone (Modifier)
Nucleoside TypeNone (Modifier)
Product TypeTET™ Dye CEP
Quantity50 mg
Reagent TypePhosphoramidite
Shipping ConditionDry Ice
Product LineTET
Unit SizeEach
Contents & Storage
This reagent should be stored dessicated at -20°C.