1-Butanesulfonyl chloride, 98%
1-Butanesulfonyl chloride, 98%
1-Butanesulfonyl chloride, 98%
Thermo Scientific Chemicals

1-Butanesulfonyl chloride, 98%

CAS: 2386-60-9 | C4H9ClO2S | 156.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 403321000
Price (EUR)
394,00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
394,00
Each
Chemical Identifiers
CAS2386-60-9
IUPAC Namebutane-1-sulfonyl chloride
Molecular FormulaC4H9ClO2S
InChI KeyWEDIIKBPDQQQJU-UHFFFAOYSA-N
SMILESCCCCS(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to light yellow
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.4540 to 1.4570 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only