2-Butanone oxime, 99%
2-Butanone oxime, 99%
2-Butanone oxime, 99%
Thermo Scientific Chemicals

2-Butanone oxime, 99%

CAS: 96-29-7 | C4H9NO | 87.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 kg
Catalog number 403330010
Price (USD)
-
Quantity:
1 kg
Request bulk or custom format
Chemical Identifiers
CAS96-29-7
IUPAC Name(E)-N-(butan-2-ylidene)hydroxylamine
Molecular FormulaC4H9NO
InChI KeyWHIVNJATOVLWBW-SNAWJCMRSA-N
SMILESCC\C(C)=N\O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=98.5 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4410 to 1.4440 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only