N-tert-Butylacrylamide, 97%, pure
N-tert-Butylacrylamide, 97%, pure
N-tert-Butylacrylamide, 97%, pure
Thermo Scientific Chemicals

N-tert-Butylacrylamide, 97%, pure

CAS: 107-58-4 | C7H13NO | 127.19 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
產品號碼 403455000
價格 (TWD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS107-58-4
IUPAC NameN-tert-butylprop-2-enamide
Molecular FormulaC7H13NO
InChI KeyXFHJDMUEHUHAJW-UHFFFAOYSA-N
SMILESCC(C)(C)NC(=O)C=C
檢視更多
規格Specification Sheet規格表
Appearance (Color)White to off-white
Infrared spectrumConforms
Melting point126°C to 131°C
Appearance (Form)Powder and/or chunks
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only