Calcium carbonate, ACS reagent, chelometric standard
Calcium carbonate, ACS reagent, chelometric standard
Calcium carbonate, ACS reagent, chelometric standard
Thermo Scientific Chemicals

Calcium carbonate, ACS reagent, chelometric standard

CAS: 471-34-1 | CCaO3 | 100.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 403801000
Price (USD)
37.65
Special offer
Online exclusive
Ends: 28-Dec-2026
44.20
Save 6.55
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
37.65
Special offer
Online exclusive
Ends: 28-Dec-2026
44.20
Save 6.55
Each
Chemical Identifiers
CAS471-34-1
IUPAC Namecalcium carbonate
Molecular FormulaCCaO3
InChI KeyVTYYLEPIZMXCLO-UHFFFAOYSA-L
SMILES[Ca++].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Titration Complexometric99.95 to 100.05 % (on dried substance)
Insoluble matter=<0.01 % (in dilute HCl)
Chloride (Cl)=<0.001 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only