Calcium carbonate, 98%, pure, light powder
Calcium carbonate, 98%, pure, light powder
Calcium carbonate, 98%, pure, light powder
Thermo Scientific Chemicals

Calcium carbonate, 98%, pure, light powder

CAS: 471-34-1 | CCaO3 | 100.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number 403815000
Price (USD)
42.70
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
42.70
Each
Chemical Identifiers
CAS471-34-1
IUPAC Namecalcium carbonate
Molecular FormulaCCaO3
InChI KeyVTYYLEPIZMXCLO-UHFFFAOYSA-L
SMILES[Ca++].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Titration Complexometric>=97.5 %
Loss on drying=<2 % (200°C, 4 h)
Acid insoluble substances=<0.2 %
Arsenic (As)=<3 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only