5(6)-Carboxifluoresceína
5(6)-Carboxifluoresceína
5(6)-Carboxifluoresceína
Thermo Scientific Chemicals

5(6)-Carboxifluoresceína

CAS: 72088-94-9 | C42H24O14 | 752.64 g/mol
Have Questions?
Número de catálogoCantidadEnvase
404105000500 mgFrasco de vidrio
Número de catálogo 404105000
Precio (USD)
-
Cantidad:
500 mg
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS72088-94-9
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)Orange to brown
Infrared spectrumConforms
Appearance (Form)Crystalline powder and/or chunks
Molar absorption coefficient>=72000 l/(mol.cm) (at Lamda max 492 nm, in 0.01N NaOH)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only