Carmine, pure, certified, alum lake of carminic acid
Carmine, pure, certified, alum lake of carminic acid
Carmine, pure, certified, alum lake of carminic acid
Thermo Scientific Chemicals

Carmine, pure, certified, alum lake of carminic acid

CAS: 1390-65-4 | C22H20O13 | 492.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 404141000
Price (USD)
449.65
Special offer
Online exclusive
Ends: 28-Dec-2026
529.00
Save 79.35 (15%)
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
449.65
Special offer
Online exclusive
Ends: 28-Dec-2026
529.00
Save 79.35 (15%)
Each
Chemical Identifiers
CAS1390-65-4
IUPAC Name3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo-7-[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]-9,10-dihydroanthracene-2-carboxylic acid
Molecular FormulaC22H20O13
InChI KeyDGQLVPJVXFOQEV-UHFFFAOYNA-N
SMILESCC1=C(C(O)=O)C(O)=CC2=C1C(=O)C1=C(O)C(C3OC(CO)C(O)C(O)C3O)=C(O)C(O)=C1C2=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
UV spectrumComplies
Appearance (Form)Powder
BSC CertificationPasses
Appearance (Color)Dark red
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only