Cellulose Acetate Butyrate, Butyryl Content 50-54%
Cellulose Acetate Butyrate, Butyryl Content 50-54%
Cellulose Acetate Butyrate, Butyryl Content 50-54%
Thermo Scientific Chemicals

Cellulose Acetate Butyrate, Butyryl Content 50-54%

CAS: 9004-36-8 | (C12H14O10)n
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 404225000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS9004-36-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline or granular powder
Acetyl content=<4 %
Hydroxyl content1.5 % (typical)
Average molecular weight16000 (Mn)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only