chlorodifluoroacetic acid sodium salt, 96%
chlorodifluoroacetic acid sodium salt, 96%
chlorodifluoroacetic acid sodium salt, 96%
Thermo Scientific Chemicals

chlorodifluoroacetic acid sodium salt, 96%

CAS: 1895-39-2 | C2ClF2NaO2 | 152.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 404591000
Price (USD)
705.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
705.00
Each
Chemical Identifiers
CAS1895-39-2
IUPAC Namesodium 2-chloro-2,2-difluoroacetate
Molecular FormulaC2ClF2NaO2
InChI KeyMRTAVLDNYYEJHK-UHFFFAOYSA-M
SMILES[Na+].[O-]C(=O)C(F)(F)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with HClO4>=95 %
Appearance (Color)White to off-white
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only