1-Chlorohexane, 95%
1-Chlorohexane, 95%
1-Chlorohexane, 95%
Acros Organics

1-Chlorohexane, 95%

CAS: 544-10-5 | C6H13Cl | 120.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Packaging:
Glass bottle
Catalog number 404645000
HSN 98020000
Price (INR)
26,077.00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
26,077.00
Each
Chemical Identifiers
CAS544-10-5
IUPAC Name1-chlorohexane
Molecular FormulaC6H13Cl
InChI KeyMLRVZFYXUZQSRU-UHFFFAOYSA-N
SMILESCCCCCCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
GC>=94 %
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.4190 to 1.4210 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only