4-Chlorophenylhydrazine hydrochloride, 97%
4-Chlorophenylhydrazine hydrochloride, 97%
4-Chlorophenylhydrazine hydrochloride, 97%
Thermo Scientific Chemicals

4-Chlorophenylhydrazine hydrochloride, 97%

CAS: 1073-70-7 | C6H7ClN2·HCl | 179.05 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) 404951000
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1073-70-7
IUPAC Namehydrogen (4-chlorophenyl)hydrazine chloride
Molecular FormulaC6H8Cl2N2
InChI KeyYQVZREHUWCCHHX-UHFFFAOYSA-N
SMILES[H+].[Cl-].NNC1=CC=C(Cl)C=C1
さらに表示
Appearance (Color)White to yellow to beige to pink to light brown
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=96.0 %
Water=<3 % (K.F.)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only