xMAP™ Sheath Concentrate PLUS, RUO
Invitrogen™
xMAP™ Sheath Concentrate PLUS, RUO
xMAP Sheath Concentrate PLUS, RUO, is the delivery medium that carries the sample to the optics component of Luminex xMAP technology-based instruments.
Have Questions?
Catalog NumberQuantity
40500231 L
Catalog number 4050023
Price (USD)
92.00
Each
Quantity:
1 L
Price (USD)
92.00
Each
xMAP Sheath Concentrate PLUS, RUO, is the delivery medium that carries the sample to the optics component of Luminex xMAP technology-based instruments. It is needed to operate all Luminex 200, FLEXMAP 3D, xMAP INTELLIFLEX, and xMAP INTELLIFLEX DRSE systems.
  • 175 mL replacement bottle for wash solution
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Recommended StorageStore at 15°C to 30°C.
Concentration20X
For Use With (Equipment)Luminex 200 Instrument System, Luminex FLEXMAP 3 D Instrument System, Luminex xMAP INTELLIFLEX, Luminex xMAP INTELLIFLEX DR-SE
Quantity1 L
Unit SizeEach
Frequently asked questions (FAQs)
What is the difference between xMAP Sheath Concentrate PLUS, RUO and xMAP Sheath Fluid Plus?
xMAP Sheath Concentrate PLUS, RUO (Cat. No. 4050023) is a 20X concentrated version of xMAP Sheath Fluid Plus (Cat. No. 4050021). It needs to be diluted before use.
xMAP Sheath Concentrate PLUS, RUO (Cat. No. 4050023) is a reformulation of xMAP Sheath Fluid Concentrate (Cat. No. 4050018). The formulation is updated from a single preservative to a mixture of three preservatives to allow for greater transit stability and to reduce the likelihood of future acquired microbial resistance.