Benzyl chloride, 99%, pure, stabilized
Benzyl chloride, 99%, pure, stabilized
Benzyl chloride, 99%, pure, stabilized
Thermo Scientific Chemicals

Benzyl chloride, 99%, pure, stabilized

CAS: 100-44-7 | C7H7Cl | 126.583 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
1 kg
2.5 kg
Packaging:
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 405090010
Price (EUR)
-
Quantity:
1 kg
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS100-44-7
IUPAC Name(chloromethyl)benzene
Molecular FormulaC7H7Cl
InChI KeyKCXMKQUNVWSEMD-UHFFFAOYSA-N
SMILESClCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Stabilizer0.25 % propylene oxide
Infrared spectrumConforms
Refractive index1.5370 to 1.5390 (20°C, 589 nm)
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
RUO – Research Use Only