Cobaltocene, 98%
Cobaltocene, 98%
Cobaltocene, 98%
Thermo Scientific Chemicals

Cobaltocene, 98%

CAS: 1277-43-6 | C10H10Co | 189.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Packaging:
Glass bottle
Catalog number 405320010
Price (USD)
75.40
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
75.40
Each
Chemical Identifiers
CAS1277-43-6
IUPAC Nameλ²-cobalt(2+) bis(cyclopenta-2,4-dien-1-ide)
Molecular FormulaC10H10Co
InChI KeyILZSSCVGGYJLOG-UHFFFAOYSA-N
SMILES[Co++].[CH-]1C=CC=C1.[CH-]1C=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Purple to black
Infrared spectrumConforms
Appearance (Form)Crystalline powder or crystals and/or chunks
Elemental Analysis>=62.0 % (C)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only