Cresyl Violet acetate, pure, certified
Cresyl Violet acetate, pure, certified
Cresyl Violet acetate, pure, certified
Thermo Scientific Chemicals

Cresyl Violet acetate, pure, certified

CAS: 10510-54-0 | C18H15N3O3 | 321.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 g
10 g
50 g
Packaging:
Glass bottle
Catalog number 405760500
Price (USD)
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS10510-54-0
IUPAC Name8,12-diamino-10λ⁴-oxa-5-azatetraphen-10-ylium acetate
Molecular FormulaC18H15N3O3
InChI KeyXKOCOMKJPWEOHX-UHFFFAOYSA-M
SMILESCC([O-])=O.NC1=CC2=[O+]C3=C(N=C2C=C1)C1=CC=CC=C1C(N)=C3
View more
SpecificationsSpecification SheetSpecification Sheet
Dye content>=65 %
Absorption Ratio P-15/P+150.94 to 1.2
Appearance (Color)Dark green to black
BSC CertificationPasses
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only