Cyclohexyldiphenylphosphine, 98%
Cyclohexyldiphenylphosphine, 98%
Cyclohexyldiphenylphosphine, 98%
Thermo Scientific Chemicals

Cyclohexyldiphenylphosphine, 98%

CAS: 6372-42-5 | C18H21P | 268.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 406100010
Price (USD)
33.65
Special offer
Online exclusive
Ends: 28-Dec-2026
39.50
Save 5.85
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
33.65
Special offer
Online exclusive
Ends: 28-Dec-2026
39.50
Save 5.85
Each
Chemical Identifiers
CAS6372-42-5
IUPAC Namecyclohexyldiphenylphosphane
Molecular FormulaC18H21P
InChI KeyZXKWUYWWVSKKQZ-UHFFFAOYSA-N
SMILESC1CCC(CC1)P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
Titration with HClO4>=97.5 %
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only