L-Cystine, specified according to the requirements of Ph.Eur.
L-Cystine, specified according to the requirements of Ph.Eur.
L-Cystine, specified according to the requirements of Ph.Eur.
Acros Organics

L-Cystine, specified according to the requirements of Ph.Eur.

CAS: 56-89-3 | C6H12N2O4S2 | 240.292 g/mol
Have Questions?
Catalog NumberQuantity
40616025025 g
Catalog number 406160250
HSN 29309040
Price (INR)
5,969.00
Each
Quantity:
25 g
Request bulk or custom format
Price (INR)
5,969.00
Each
Chemical Identifiers
CAS56-89-3
IUPAC Name(2R)-2-amino-3-{[(2R)-2-amino-2-carboxyethyl]disulfanyl}propanoic acid
Molecular FormulaC6H12N2O4S2
InChI KeyLEVWYRKDKASIDU-IMJSIDKUSA-N
SMILESN[C@@H](CSSC[C@H](N)C(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Specific optical rotationOn dried substance
Iron (Fe)=<10 ppm
Appearance of solutionthan reference solution Y7
Ninhydrin-positive substancesPasses test
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only