Decahydronaphthalene, 99%, mixture of cis and trans, anhydrous, AcroSeal™
Decahydronaphthalene, 99%, mixture of cis and trans, anhydrous, AcroSeal™
Decahydronaphthalene, 99%, mixture of cis and trans, anhydrous, AcroSeal™
Decahydronaphthalene, 99%, mixture of cis and trans, anhydrous, AcroSeal™
Thermo Scientific Chemicals

Decahydronaphthalene, 99%, mixture of cis and trans, anhydrous, AcroSeal™

CAS: 91-17-8 | C10H18 | 138.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number 406171000
Price (USD)
-
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS91-17-8
IUPAC Namedecahydronaphthalene
Molecular FormulaC10H18
InChI KeyNNBZCPXTIHJBJL-UHFFFAOYSA-N
SMILESC1CCC2CCCCC2C1
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.005 %
Infrared spectrumConforms
Appearance (Form)Clear liquid
GC>=98.5 % (mixture of cis and trans)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only