Thermo Scientific™
Miscellaneous Microtome Accessories
Related applications:
Anatomical Pathology
Support and expand the functionality of Thermo Scientific™ 200 and 300 series of microtomes with Thermo Scientific Miscellaneous Microtome Accessories.
Back to top
  • Interlock plug replaces foot pedal connector
  • Storage unit can be removed and used separately; tray for specimens can be cooled by changeable inserts
  • Large-field magnifier attaches to facilitate knife alignment and approach for sectioning; magnification 2.5X