4,4'-Diaminostilbene-2,2'-disulfonic acid, 95%
4,4'-Diaminostilbene-2,2'-disulfonic acid, 95%
4,4'-Diaminostilbene-2,2'-disulfonic acid, 95%
Thermo Scientific Chemicals

4,4'-Diaminostilbene-2,2'-disulfonic acid, 95%

CAS: 81-11-8 | C14H14N2O6S2 | 370.4 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 406415000
Price (USD)
320.00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
320.00
Each
Chemical Identifiers
CAS81-11-8
IUPAC Name5-amino-2-[(1E)-2-(4-amino-2-sulfophenyl)ethenyl]benzene-1-sulfonic acid
Molecular FormulaC14H14N2O6S2
InChI KeyREJHVSOVQBJEBF-OWOJBTEDSA-N
SMILESNC1=CC=C(\C=C\C2=CC=C(N)C=C2S(O)(=O)=O)C(=C1)S(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to yellow-brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with NaOH>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only