trans-1,4-Dibromo-2-butene, 97%
trans-1,4-Dibromo-2-butene, 97%
trans-1,4-Dibromo-2-butene, 97%
Thermo Scientific Chemicals

trans-1,4-Dibromo-2-butene, 97%

CAS: 821-06-7 | C4H6Br2 | 213.9 g/mol
製品番号(カタログ番号)数量パッケージ
406461000100 gGlass bottle
製品番号(カタログ番号) 406461000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS821-06-7
IUPAC Name(2E)-1,4-dibromobut-2-ene
Molecular FormulaC4H6Br2
InChI KeyRMXLHIUHKIVPAB-OWOJBTEDSA-N
SMILESBrCC=CCBr
さらに表示
Appearance (Color)White to beige to brown
Appearance (Form)Crystalline powder or crystals
GC>=96.0 %
Infrared spectrumConforms
Melting point48°C to 53°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only