1,2-Dibromo-2-methylpropane, 98%
1,2-Dibromo-2-methylpropane, 98%
1,2-Dibromo-2-methylpropane, 98%
Thermo Scientific Chemicals

1,2-Dibromo-2-methylpropane, 98%

CAS: 594-34-3 | C4H8Br2 | 215.92 g/mol
数量:
5 g
25 g
250 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 406492500
価格(JPY)
-
数量:
250 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS594-34-3
IUPAC Name1,2-dibromo-2-methylpropane
Molecular FormulaC4H8Br2
InChI KeySDTXSEXYPROZSZ-UHFFFAOYSA-N
SMILESCC(C)(Br)CBr
さらに表示
Infrared spectrumConforms
GC>=97.5 %
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Refractive index1.5083 to 1.5103 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only