1,2-Dichloroethane, 99+%, ACS reagent, meets the requirements of Reag. Ph.Eur.
1,2-Dichloroethane, 99+%, ACS reagent, meets the requirements of Reag. Ph.Eur.
1,2-Dichloroethane, 99+%, ACS reagent, meets the requirements of Reag. Ph.Eur.
Thermo Scientific Chemicals

1,2-Dichloroethane, 99+%, ACS reagent, meets the requirements of Reag. Ph.Eur.

CAS: 107-06-2 | C2H4Cl2 | 98.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 406820010
Price (USD)
-
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS107-06-2
IUPAC Name1,2-dichloroethane
Molecular FormulaC2H4Cl2
InChI KeyWSLDOOZREJYCGB-UHFFFAOYSA-N
SMILESClCCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Density(20/20°C) about 1.25
Color scale=<10 APHA
Water=<0.03 % (Coulometric)
Titrable acid=<0.0003 meq/g
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only