Diethyl adipate, 99%
Diethyl adipate, 99%
Diethyl adipate, 99%
Thermo Scientific Chemicals

Diethyl adipate, 99%

CAS: 141-28-6 | C10H18O4 | 202.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 407171000
Price (USD)
20.65
Special offer
Online exclusive
Ends: 28-Dec-2026
24.00
Save 3.35
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
20.65
Special offer
Online exclusive
Ends: 28-Dec-2026
24.00
Save 3.35
Each
Chemical Identifiers
CAS141-28-6
IUPAC Name1,6-diethyl hexanedioate
Molecular FormulaC10H18O4
InChI KeyVIZORQUEIQEFRT-UHFFFAOYSA-N
SMILESCCOC(=O)CCCCC(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Refractive index1.4260 to 1.4280 (20°C, 589 nm)
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only