Diethylenetriaminepentaacetic acid, pentasodium salt, tech., 40% aqueous solution
Diethylenetriaminepentaacetic acid, pentasodium salt, tech., 40% aqueous solution
Diethylenetriaminepentaacetic acid, pentasodium salt, tech., 40% aqueous solution
Thermo Scientific Chemicals

Diethylenetriaminepentaacetic acid, pentasodium salt, tech., 40% aqueous solution

CAS: 140-01-2 | C14H18N3Na5O10 | 503.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
3 kg
Packaging:
Glass bottle
Catalog number 407290010
Price (USD)
142.65
Special offer
Online exclusive
Ends: 28-Dec-2026
168.00
Save 25.35
Each
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
142.65
Special offer
Online exclusive
Ends: 28-Dec-2026
168.00
Save 25.35
Each
Chemical Identifiers
CAS140-01-2
SpecificationsSpecification SheetSpecification Sheet
Color scale=<150 APHA
pH11.0 to 12.0 (1 % in water)
Appearance (Form)Clear solution
Titration39 to 41 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only