Diethyl terephthalate, 98%
Diethyl terephthalate, 98%
Diethyl terephthalate, 98%
Thermo Scientific Chemicals

Diethyl terephthalate, 98%

CAS: 636-09-9 | C12H14O4 | 222.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 407450250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS636-09-9
IUPAC Name1,4-diethyl benzene-1,4-dicarboxylate
Molecular FormulaC12H14O4
InChI KeyONIHPYYWNBVMID-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CC=C(C=C1)C(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=97.5 %
Appearance (Color)White to cream
Appearance (Form)Powder
Melting point43°C to 47°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only